ENA187623021 Enamine

2-methyl-3-(1,3,4-oxadiazol-2-yl)aniline

  • MDL number MFCD08444030
  • Cas Number : 924869-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85...87
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42409

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42409
Availability details for
Quantity
  • Availability
  • Add to Cart