ENA187623035 Enamine

N-[2-(thiophen-2-yl)ethyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxamide

  • MDL number MFCD09932283
  • Cas Number : 1101919-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42439
Availability details for
Quantity
  • Availability
  • Add to Cart