ENA187623039 Enamine

N-[2-(4-fluorophenyl)ethyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxamide

  • MDL number MFCD09935805
  • Cas Number : 1105059-12-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42445

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42445
Availability details for
Quantity
  • Availability
  • Add to Cart