ENA187623041 Enamine

[6-(azepan-1-yl)pyridin-3-yl]methanamine

  • MDL number MFCD09045371
  • Cas Number : 926204-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42449

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42449
Availability details for
Quantity
  • Availability
  • Add to Cart