ENA187623079 Enamine

N-(3-fluorophenyl)-2-(piperazin-1-yl)propanamide

  • MDL number MFCD10011294
  • Cas Number : 1018252-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42542

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42542
Availability details for
Quantity
  • Availability
  • Add to Cart