ENA187623081 Enamine

2,2-dimethylpropanohydrazide

  • MDL number MFCD01310839
  • Cas Number : 42826-42-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-41553

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-41553
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart