ENA187623091 Enamine

N'-hydroxy-2-(propan-2-yloxy)ethanimidamide

  • Cas Number : 1016754-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42566

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42566
Availability details for
Quantity
  • Availability
  • Add to Cart