ENA187623101 Enamine

N-{1-[(pyridin-2-yl)methyl]piperidin-4-ylidene}hydroxylamine

  • MDL number MFCD09942508
  • Cas Number : 1016869-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42587
Availability details for
Quantity
  • Availability
  • Add to Cart