ENA187623109 Enamine

2-{4-[(pyridin-2-yl)methoxy]phenyl}acetonitrile

  • MDL number MFCD09950290
  • Cas Number : 1017037-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42597
Availability details for
Quantity
  • Availability
  • Add to Cart