ENA187623111 Enamine

1-{3-[(propan-2-yloxy)methyl]phenyl}methanamine

  • MDL number MFCD09805148
  • Cas Number : 1016717-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42603
Availability details for
Quantity
  • Availability
  • Add to Cart