ENA187623112 Enamine

[1-(4-fluorophenyl)cyclopropyl]methanamine

  • MDL number MFCD06213570
  • Cas Number : 75180-46-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42604

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42604
Availability details for
Quantity
  • Availability
  • Add to Cart