ENA187623113 Enamine

2-[(benzyloxy)methyl]aniline

  • MDL number MFCD09814533
  • Cas Number : 1016498-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42605
Availability details for
Quantity
  • Availability
  • Add to Cart