ENA187623118 Enamine

4-(piperidin-4-yl)phenol hydrochloride

  • MDL number MFCD10565007
  • Cas Number : 263139-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-30670

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-30670
Availability details for
Quantity
  • Availability
  • Add to Cart