ENA187623122 Enamine

2-{[(2-nitrophenyl)methyl]sulfanyl}pyrimidine

  • MDL number MFCD11857929
  • Cas Number : 1118787-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42623

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42623
Availability details for
Quantity
  • Availability
  • Add to Cart