ENA187623128 Enamine

4-(2-methoxyethoxy)-3-nitrobenzoic acid

  • MDL number MFCD09808418
  • Cas Number : 90923-45-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42632

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42632
Availability details for
Quantity
  • Availability
  • Add to Cart