ENA187623131 Enamine

7-oxo-5-phenyl-4H,7H-[1,2,4]triazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD04628465
  • Cas Number : 329929-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42635
Availability details for
Quantity
  • Availability
  • Add to Cart