ENA187623132 Enamine

ethyl 2-[4-(benzenesulfonyl)piperazin-1-yl]acetate

  • MDL number MFCD06052956
  • Cas Number : 890281-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-41879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41879
Availability details for
Quantity
  • Availability
  • Add to Cart