ENA187623135 Enamine

4-[(cyclopropylamino)methyl]phenol

  • MDL number MFCD09048706
  • Cas Number : 926201-89-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42639
Availability details for
Quantity
  • Availability
  • Add to Cart