ENA187623146 Enamine

ethyl 2-(4-cyano-2-methoxyphenoxy)acetate

  • MDL number MFCD02256122
  • Cas Number : 445014-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42670

Safety Information

msds https://www.enaminestore.com/catalog/EN300-42670
Availability details for
Quantity
  • Availability
  • Add to Cart