ENA187623156 Enamine

N-methyl-1-(5-methyl-1,2-oxazol-3-yl)methanesulfonamide

  • MDL number MFCD11172477
  • Cas Number : 1042787-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42709

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42709
Availability details for
Quantity
  • Availability
  • Add to Cart