ENA187623158 Enamine

1-benzyl-1-methylurea

  • MDL number MFCD09811057
  • Cas Number : 73355-62-1
  • Molecular Weight : 164.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42713

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42713
Availability details for
Quantity
  • Availability
  • Add to Cart