ENA187623162 Enamine

2-{[(2-fluorophenyl)carbamoyl]amino}acetic acid

  • MDL number MFCD02112804
  • Cas Number : 387882-09-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-42717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42717
Availability details for
Quantity
  • Availability
  • Add to Cart