ENA187623163 Enamine

N-(6-amino-1,3-benzothiazol-2-yl)-5-methylthiophene-2-carboxamide

  • MDL number MFCD09805280
  • Cas Number : 1016519-15-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-42718

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42718
Availability details for
Quantity
  • Availability
  • Add to Cart