ENA187623170 Enamine

2-{[(2-chlorophenyl)methyl]amino}propan-1-ol

  • MDL number MFCD11840121
  • Cas Number : 1118787-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42015
Availability details for
Quantity
  • Availability
  • Add to Cart