ENA187623172 Enamine

3-methyl-1-(2-methylpropyl)-2,4-dioxo-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-6-carboxylic acid

  • MDL number MFCD09051867
  • Cas Number : 937624-04-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42027

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42027
Availability details for
Quantity
  • Availability
  • Add to Cart