ENA187623173 Enamine

2-chloro-N-cyclopropyl-5-nitrobenzamide

  • MDL number MFCD03367842
  • Cas Number : 83909-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42029

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42029
Availability details for
Quantity
  • Availability
  • Add to Cart