ENA187623181 Enamine

6-(2-methyl-1H-imidazol-1-yl)pyridin-3-amine

  • MDL number MFCD08700227
  • Cas Number : 430535-32-3
  • Molecular Weight : 174.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42033
Availability details for
Quantity
  • Availability
  • Add to Cart