ENA187623186 Enamine

2-(3-phenyl-1H-1,2,4-triazol-5-yl)aniline

  • MDL number MFCD11857869
  • Molecular Weight : 236.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-42040

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42040
Availability details for
Quantity
  • Availability
  • Add to Cart