ENA187623187 Enamine

2-(dimethylamino)-N'-hydroxyethanimidamide

  • MDL number MFCD05663028
  • Cas Number : 67015-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 135...137
Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42041

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-42041
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart