ENA187623190 Enamine

4-(2,5-difluorobenzoyl)piperidine hydrochloride

  • MDL number MFCD11812209
  • Cas Number : 1172297-96-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-42044

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-42044
Availability details for
Quantity
  • Availability
  • Add to Cart