ENA189566328 Enamine

2-amino-3-(3-hydroxy-5-methoxyphenyl)propanoic acid

  • MDL number MFCD11858020
  • Cas Number : 91819-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43155
Availability details for
Quantity
  • Availability
  • Add to Cart