ENA189566330 Enamine

4-(1,3-thiazol-2-yl)aniline dihydrochloride

  • MDL number MFCD11858036
  • Cas Number : 1181458-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43235
Availability details for
Quantity
  • Availability
  • Add to Cart