ENA189566336 Enamine

1-(pyrimidin-2-yl)piperidin-3-amine

  • MDL number MFCD11858051
  • Cas Number : 1146290-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43309

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43309
Availability details for
Quantity
  • Availability
  • Add to Cart