ENA189566343 Enamine

2-fluoro-4-nitrobenzene-1-sulfonyl chloride

  • MDL number MFCD11858095
  • Cas Number : 1146290-36-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69-70
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43514

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43514
Availability details for
Quantity
  • Availability
  • Add to Cart