ENA189566345 Enamine

7-chloro-5-(chloromethyl)-2,2-dimethyl-2,3-dihydro-1-benzofuran

  • MDL number MFCD11858107
  • Cas Number : 58023-29-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43554
Availability details for
Quantity
  • Availability
  • Add to Cart