ENA189566348 Enamine

N-(2-aminoethyl)cyclohexanecarboxamide hydrochloride

  • MDL number MFCD11858112
  • Cas Number : 1181458-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43604
Availability details for
Quantity
  • Availability
  • Add to Cart