ENA189566400 Enamine

{3-[3-(4-fluorophenyl)-1H-pyrazol-5-yl]propyl}(methyl)amine

  • MDL number MFCD11858013
  • Cas Number : 1192579-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43128

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43128
Availability details for
Quantity
  • Availability
  • Add to Cart