ENA189566401 Enamine

2-(piperidin-3-yl)-1H-indole

  • MDL number MFCD05207657
  • Cas Number : 92960-62-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 175...177
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43130

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-43130
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart