ENA189566404 Enamine

tert-butyl N-[2-(5-amino-4-cyano-1H-1,2,3-triazol-1-yl)ethyl]carbamate

  • MDL number MFCD09702268
  • Cas Number : 929975-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43135

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43135
Availability details for
Quantity
  • Availability
  • Add to Cart