ENA189566405 Enamine

1-phenyl-5-(pyridin-3-yl)-1H-pyrazole-3-carboxylic acid hydrochloride

  • MDL number MFCD11858015
  • Cas Number : 1172372-17-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43136
Availability details for
Quantity
  • Availability
  • Add to Cart