ENA189566413 Enamine

3-methyl-1,2,4-oxadiazol-5-amine

  • MDL number MFCD08272076
  • Cas Number : 3663-39-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 159-161
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-33308

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-33308
Availability details for
Quantity
  • Availability
  • Add to Cart