ENA189566415 Enamine

3,4-dihydro-2H-pyran-5-carbaldehyde

  • MDL number MFCD00085019
  • Cas Number : 25090-33-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43151

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43151
Availability details for
Quantity
  • Availability
  • Add to Cart