ENA189566421 Enamine

4-(4-bromobenzenesulfonyl)aniline hydrochloride

  • MDL number MFCD11858023
  • Cas Number : 1172364-06-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43160
Availability details for
Quantity
  • Availability
  • Add to Cart