ENA189566424 Enamine

1-[6-(4-fluorophenoxy)pyridin-3-yl]methanamine

  • MDL number MFCD09732743
  • Cas Number : 620608-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 46-48
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43164

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43164
Availability details for
Quantity
  • Availability
  • Add to Cart