ENA189566429 Enamine

1-(4-butoxy-3-methoxyphenyl)ethan-1-amine hydrochloride

  • MDL number MFCD11858026
  • Cas Number : 1171786-31-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-43171
Availability details for
Quantity
  • Availability
  • Add to Cart