ENA189566436 Enamine

3-amino-4-fluorobenzene-1-sulfonamide

  • MDL number MFCD09900531
  • Cas Number : 1017448-36-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43190
Availability details for
Quantity
  • Availability
  • Add to Cart