ENA189566449 Enamine

3,5-dimethyl-1-(piperidin-4-yl)piperidine dihydrochloride

  • MDL number MFCD11858032
  • Cas Number : 1181458-51-4
  • Molecular Weight : 269.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43207

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43207
Availability details for
Quantity
  • Availability
  • Add to Cart