ENA189566460 Enamine

2-(4-methylpiperazine-1-carbonyl)-1,2,3,4-tetrahydroquinoline

  • MDL number MFCD09936376
  • Cas Number : 1104561-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43221

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43221
Availability details for
Quantity
  • Availability
  • Add to Cart