ENA189566468 Enamine

N-methylpyrimidin-2-amine

  • MDL number MFCD00234126
  • Cas Number : 931-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 58-62
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-43236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43236
Availability details for
Quantity
  • Availability
  • Add to Cart