ENA189566469 Enamine

2,6-dimethyl-4-(piperidin-4-yl)morpholine dihydrochloride

  • MDL number MFCD08448215
  • Cas Number : 1181457-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-43238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-43238
Availability details for
Quantity
  • Availability
  • Add to Cart